Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGI-62 Monoclonal Antibody (OTI4B5)
GREENER_CHOICE

Catalog No. PIMA526080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526080 Supplier Invitrogen™ Supplier No. MA526080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CGI-62 Monoclonal Antibody for Western Blot, IHC (P)

The CGI-62 gene encodes a hypothetical protein located on chromosome 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGI-62
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZC2HC1A
Gene Accession No. Q96GY0
Gene Alias 3110050N22Rik; AI790358; AU023959; C14H8ORF70; C8orf70; CGI-62; Fam164a; family with sequence similarity 164, member A; protein FAM164A; RGD1311970; UPF0418 protein FAM164A; Zc2hc1a; Zinc finger C2HC domain-containing protein 1A; zinc finger C2HC-type containing 1A; zinc finger, C2HC-type containing 1A
Gene Symbols ZC2HC1A
Host Species Mouse
Immunogen Full length human recombinant protein of FAM164A produced in E.coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51101
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.