Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGI58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518666
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518666 Supplier Invitrogen™ Supplier No. PA518666
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGI58
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABHD5
Gene Accession No. Q8WTS1, Q9DBL9
Gene Alias 1300003D03Rik; 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 2010002J10Rik; ABHD5; abhydrolase domain containing 5; abhydrolase domain-containing protein 5; aCGI58; CDS; CGI 58; CGI58; CGI-58; CGI58 protein; IECN2; IECN5; lipid droplet-binding protein CGI-58; MGC8731; NCIE2; Protein CGI-58; truncated abhydrolase domain-containing protein 5
Gene Symbols ABHD5
Host Species Goat
Immunogen Synthetic peptide sequence (FPERPDLADQDR) corresponding to the internal amino acids of ABHD5 (aa 181-192).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51099, 67469
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.