Learn More
Invitrogen™ CGI58 Polyclonal Antibody

Description
Peptide sequence: SVIFGARSCI DGNSGTSIQS LRPHSYVKTI AILGAGHYVY ADQPEEFNQK Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%.
Specifications
Specifications
| Antigen | CGI58 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ABHD5 |
| Gene Accession No. | Q8WTS1 |
| Gene Alias | 1300003D03Rik; 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 2010002J10Rik; ABHD5; abhydrolase domain containing 5; abhydrolase domain-containing protein 5; aCGI58; CDS; CGI 58; CGI58; CGI-58; CGI58 protein; IECN2; IECN5; lipid droplet-binding protein CGI-58; MGC8731; NCIE2; Protein CGI-58; truncated abhydrolase domain-containing protein 5 |
| Gene Symbols | ABHD5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.