Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cGK2/PRKG2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305940DL594
Description
cGK2/PRKG2 Polyclonal specifically detects cGK2/PRKG2 in Human samples. It is validated for Western Blot.Specifications
cGK2/PRKG2 | |
Polyclonal | |
Western Blot | |
cGK 2, cGKIIcGK2, cGMP-dependent protein kinase 2, cGMP-dependent protein kinase II, EC 2.7.11, PRKGR2EC 2.7.11.12, protein kinase, cGMP-dependent, type II | |
Produced in rabbits immunized with E. coli-derived Human cGK2/PRKG2 fragment. | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
5593 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction