Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cGKI Monoclonal Antibody (OTI9A4)
GREENER_CHOICE

Catalog No. PIMA525110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525110 Supplier Invitrogen™ Supplier No. MA525110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cGKI Monoclonal Antibody for Western Blot, ICC/IF, Flow

PRKG1 is a cGMP-dependent serine/threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation. The PRKG1 proteins play a central role in regulating cardiovascular and neuronal functions in addition to relaxing smooth muscle tone, and modulating cell growth. This gene is most strongly expressed in all types of smooth muscle, platelets, cerebellar Purkinje cells, hippocampal neurons, and the lateral amygdala. Isoforms Ialpha and Ibeta have identical cGMP-binding and catalytic domains but differ in their leucine/isoleucine zipper and autoinhibitory sequences and therefore differ in their dimerization substrates and kinase enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cGKI
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI9A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKG1
Gene Accession No. Q13976
Gene Alias AAT8; AW125416; cGK; cGK 1; CGK 1 alpha; cGK 1 beta; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; cGMP dependent protein kinase type 1; cGMP kinase type I alpha; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase 1, alpha isozyme; cGMP-dependent protein kinase 1, beta isozyme; cGMP-dependent protein kinase I; Gm19690; OTTHUMP00000019618; PKG; Pkgi; Prkg1; PRKG1B; PRKGR1A; PRKGR1B; protein kinase cGMP-dependent 1; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type 1; protein kinase, cGMP-dependent, type I; RP11-346D6.1; Unknown (protein for MGC:134126); unnamed protein product
Gene Symbols PRKG1
Host Species Mouse
Immunogen Full length human recombinant protein of PRKG1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5592
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.