Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cGKI Monoclonal Antibody (T.344.1)
GREENER_CHOICE

Catalog No. PIMA514862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514862 Supplier Invitrogen™ Supplier No. MA514862
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with PKG-2.

PRKG1 is a cGMP-dependent serine/threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation. The PRKG1 proteins play a central role in regulating cardiovascular and neuronal functions in addition to relaxing smooth muscle tone, and modulating cell growth. This gene is most strongly expressed in all types of smooth muscle, platelets, cerebellar Purkinje cells, hippocampal neurons, and the lateral amygdala. Isoforms Ialpha and Ibeta have identical cGMP-binding and catalytic domains but differ in their leucine/isoleucine zipper and autoinhibitory sequences and therefore differ in their dimerization substrates and kinase enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cGKI
Applications Western Blot
Classification Monoclonal
Clone T.344.1
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene PRKG1
Gene Accession No. P0C605, Q13976
Gene Alias AAT8; AW125416; cGK; cGK 1; CGK 1 alpha; cGK 1 beta; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; cGMP dependent protein kinase type 1; cGMP kinase type I alpha; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase 1, alpha isozyme; cGMP-dependent protein kinase 1, beta isozyme; cGMP-dependent protein kinase I; Gm19690; OTTHUMP00000019618; PKG; Pkgi; Prkg1; PRKG1B; PRKGR1A; PRKGR1B; protein kinase cGMP-dependent 1; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type 1; protein kinase, cGMP-dependent, type I; RP11-346D6.1; Unknown (protein for MGC:134126); unnamed protein product
Gene Symbols PRKG1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to cental residues of human PKG-1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19091, 54286, 5592
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.