Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ cGKII Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116863
Description
Positive Samples: Mouse lung, Rat brain Immunogen sequence: MGNGSVKPKH SKHPDGHSGN LTTDALRNKV TELERELRRK DAEIQEREYH LKELREQLSK QTVAIAELTE ELQNKCIQLN KLQDVVHMQG GSPLQASPDK VPLEVHRKTS GLVSLHSRRG AKAGVSAEPT TRTYDLNKPP EFSFEKARVR KDSSEKKLIT.
PRKG2 (PKG2, cGKII) exhibits many cellular functions and has cAMP- and cGMP-dependent protein kinase activity. cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Identification of the pathway that mediates intestinal fluid secretion by E. coli. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.
Specifications
| cGKII | |
| Polyclonal | |
| Unconjugated | |
| PRKG2 | |
| AW212535; cGK 2; cGK2; cGKII; cGK-II; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; EC 2.7.11.12; GDPKII; PRKG2; PRKGR2; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; protein kinase, cGMP-dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19092, 25523 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2.22 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q61410, Q64595 | |
| PRKG2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human PRKG2 (NP_0062501). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction