Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531654 Supplier Invitrogen™ Supplier No. PA531654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Predicted reactivity: Human (97%), Mouse (86%), Rabbit (83%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CGN
Gene Accession No. Q9P2M7
Gene Alias 6330408J11Rik; CGN; Cingulin; KIAA1319
Gene Symbols CGN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human CGN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.