Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CGNL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548808
Description
Predicted to react with mouse based on sequence homology.
Paracingulin (CGNL1, cingulin-like 1, FLJ14957, JACOP, KIAA1749) is novel protein with similar domain organization and significant sequence homology to cingulin, one of the tight junction (TJ) molecules. Paracingulin may play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
Specifications
CGNL1 | |
Polyclonal | |
Unconjugated | |
CGNL1 | |
4933421H10Rik; 9930020M10Rik; AI503810; CGNL1; cingulin like 1; cingulin-like 1; cingulin-like protein 1; JACOP; Junction-associated coiled-coil protein; Kiaa1749; Paracingulin; PCING | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
84952 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q0VF96 | |
CGNL1 | |
KLH conjugated synthetic peptide between 317-344 amino acids from the N-terminal region of human CGNL1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction