Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563366 Supplier Invitrogen™ Supplier No. PA563366
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEKVEENSTL QQRLEESEGE LRKNLEELFQ VKMEREQHQT EIRDLQDQLS EMHDELDSAK RSEDREKGAL IEELLQAKQD LQDLLIAK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Paracingulin (CGNL1, cingulin-like 1, FLJ14957, JACOP, KIAA1749) is novel protein with similar domain organization and significant sequence homology to cingulin, one of the tight junction (TJ) molecules. Paracingulin may play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGNL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CGNL1
Gene Accession No. Q0VF96
Gene Alias 4933421H10Rik; 9930020M10Rik; AI503810; CGNL1; cingulin like 1; cingulin-like 1; cingulin-like protein 1; JACOP; Junction-associated coiled-coil protein; Kiaa1749; Paracingulin; PCING
Gene Symbols CGNL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CGNL1. Recombinant protein control fragment (Product #RP-103280).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84952
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.