Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 37157
GREENER_CHOICE

Catalog No. 111410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-141-0 10 mg
11-145-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-141-0 Supplier Tocris Bioscience™ Supplier No. 1114/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antagonist of mitochondrial Na+/Ca2+ exchange

Selective antagonist of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).

Chemical Identifiers

CAS 75450-34-9
Molecular Formula C15H11Cl2NOS

Specifications

Quantity 10 mg
Formula Weight 324.22
Purity >99%
Molecular Weight (g/mol) 324.219
Chemical Name or Material CGP 37157
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.