Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 39551, Tocris Bioscience™

Catalog No. 140950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-095-0 50 mg
14-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-095-0 Supplier R&D Systems Supplier No. 1409/50
This item is not returnable. View return policy

Potent, selective and competitive NMDA antagonist

Potent, selective and competitive NMDA antagonist (Ki = 310 nM for inhibition of [3H]-CPP binding in rat brain). Centrally active upon oral administration in vivo.

Chemical Identifiers

CAS 127910-32-1
Molecular Formula C8H16NO5P
Molecular Weight (g/mol) 237.192
InChI Key OKDOWCKDTWNRCB-GQCTYLIASA-N
Synonym 2-amino-4-methyl-5-phosphono-3 pentenoic acid carboxyethyl ester, e-+/--2-amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, 1-ethyl ester, 3e, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, 1-ethyl ester, e-+-, 2e-4-amino-5-ethoxy-2-methyl-5-oxopent-2-en-1-ylphosphonic acid, cgp hplc, e-2-amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester, e-4-amino-5-ethoxy-2-methyl-5-oxopent-2-enyl phosphonic acid
PubChem CID 6372334
IUPAC Name [(E)-4-amino-5-ethoxy-2-methyl-5-oxopent-2-enyl]phosphonic acid
SMILES CCOC(=O)C(C=C(C)CP(=O)(O)O)N

Specifications

Quantity 50 mg
Formula Weight 237.19
Percent Purity >98%
Chemical Name or Material CGP 39551
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.