Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 71683 hydrochloride
GREENER_CHOICE

Catalog No. 219910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-991-0 10 mg
21-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-991-0 Supplier Tocris Bioscience™ Supplier No. 2199/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective and potent non-peptide NPY Y5 receptor antagonist

CGP 71683 hydrochloride is an extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.

Specifications

Quantity 10 mg
Product Type CGP 71683 hydrochloride
Purity >99%
Molecular Weight (g/mol) 512.07
Inhibitors CGP 71683 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.