Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 7930
GREENER_CHOICE

Catalog No. 151310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-131-0 10 mg
15-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-131-0 Supplier Tocris Bioscience™ Supplier No. 1513/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of GABAB receptors

Positive allosteric modulator of GABAB receptors. Increases the potency and efficacy of GABA at both native and recombinant GABAB receptors (EC50 values are 5.37 and 4.60μM respectively) and enhances the inhibitory effect of the agonist L-baclofen in cultured cortical neurons. Reduces operant self-administration of ethanol in alcohol-preferring rats following i.p. administration.

Chemical Identifiers

CAS 57717-80-3
Molecular Formula C19H32O2

Specifications

Quantity 10 mg
Formula Weight 292.46
Purity >98%
Molecular Weight (g/mol) 292.463
Chemical Name or Material CGP 7930
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.