Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGR19 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299466PCP
Description
CGR19 Polyclonal antibody specifically detects CGR19 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CGR19 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
cell growth regulator with ring finger domain 1, Cell growth regulatory gene 19 protein, CGR19cell growth regulator with RING finger domain protein 1, RING finger protein 197, RNF197GCRRF1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CGR19. | |
0.1 mL | |
Cell Cycle and Replication | |
10668 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction