Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGR19 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987222
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7222 Supplier Bioss Supplier No. BS6373R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CGRRF1 is able to inhibit growth in several cell lines.

Specifications

Antigen CGR19
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CGRRF1
Gene Accession No. Q99675
Gene Alias 1110038G02Rik; 1810009H17Rik; BOS_10975; cell growth regulator with ring finger domain 1; Cell growth regulator with RING finger domain protein 1; Cell growth regulatory gene 19 protein; cell growth regulatory with ring finger domain; CGR19; CGRRF1; LOC100356220; RING finger protein 197; RNF197
Gene Symbols CGRRF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CGR19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10668
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.