Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGREF1 Antibody (001), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290214JF549
Description
CGREF1 Monoclonal antibody specifically detects CGREF1 in Human samples. It is validated for Western Blot, ELISASpecifications
CGREF1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
001 | |
Western Blot, ELISA | |
cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin | |
Rabbit immunized with purified, recombinant Human CGREF1 (Uniprot#: Q99674-1, Met1-Ile301) | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication | |
10669 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction