Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGREF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321344100UL
Description
CGREF1 Polyclonal antibody specifically detects CGREF1 in Human samples. It is validated for ImmunofluorescenceSpecifications
CGREF1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LAPSPQEPQAVGRQSLLAKSPLRQETQEAPGPREEAKGQVEARRESLDPVQEPGGQAEADGDVPGPRGEAEGQAEAKGDA | |
100 μg | |
Cell Biology, Cell Cycle and Replication | |
10669 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction