Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGREF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552536 Supplier Invitrogen™ Supplier No. PA552536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKDGVTRPDS EVQHQLLPNP FQPGQEQLGL LQSYLKGLGR TEVQLEHLSR EQVLLYLFAL HDYDQSGQLD GLELLSMLTA ALAPGAANSP TTNPVILIVD KVLETQDLNG DGLMTPAELI NFPGVALRHV EPG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.

CGREF1 (cell growth regulator with EF-hand domain 1), also known as CGR11, is a 301 amino acid secreted protein that contains two highly conserved calcium binding EF-hand domains, which are required for mediating cell-cell adhesion. Induced by p53, CGREF1 is able to inhibit cell growth in various cell lines. CGREF1 is encoded by a gene located on human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alstrom syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGREF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CGREF1
Gene Accession No. Q99674
Gene Alias 1110004G24Rik; AW045201; cell growth regulator with EF hand domain 1; cell growth regulator with EF hand domain protein 1; cell growth regulator with EF-hand domain 1; cell growth regulatory gene 11 protein; cell growth regulatory with EF-hand domain; CGR11; Cgref1; Hydrophobestin
Gene Symbols CGREF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CGREF1. Recombinant protein control fragment (Product #RP-89919).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.