Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGRP Monoclonal Antibody (5)
GREENER_CHOICE

Catalog No. 01670109
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-109 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-109 Supplier Invitrogen™ Supplier No. ABS0260502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 026-05-02 detects CGRP alpha in human and rat samples. ABS 026-05-02 binds human alpha-CGRP. Based on inhibition ELISA, this antibody shows full cross-reactivity with rat alpha-CGRP and about about 50% cross-reactivity with human beta-CGRP. Cross-reactivity with coated rat alpha-CGRP is low (8%) and there is no cross-reaction with human amylin. ABS 026-05-02 has been successfully used in ELISA and immunohistochemistry procedures.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGRP
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Calca
Gene Accession No. P01256, P01257, P01258, P06881
Gene Alias alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6
Gene Symbols Calca
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues 19-37 of human alpha-CGRP.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24241, 796
Target Species Human, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.