Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGRP Mouse anti-Human, Rat, Clone: 4, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $439.00
Specifications
Antigen | CGRP |
---|---|
Clone | 4 |
Concentration | 1.0 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CGRP Monoclonal specifically detects CGRP in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CGRP | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
796 | |
IgG1 κ | |
Protein A or G purified |
4 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat | |
alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
Synthetic human alpha-CGRP adsorbed onto aluminum hydroxide gel. The epitope lies within residues 19-37 of human alpha-CGRP. | |
Primary | |
Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title