Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGRP Mouse anti-Human, Rat, Clone: 5, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
Antigen | CGRP |
---|---|
Clone | 5 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Description
CGRP Monoclonal specifically detects CGRP in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CGRP | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human, Rat | |
alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
Synthetic human alpha-CGRP | |
Primary | |
Store at 4°C in the dark. |
5 | |
Western Blot, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
796 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title