Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGRP Recombinant Rabbit Monoclonal Antibody (BLR159J)
GREENER_CHOICE

Catalog No. PIMA544503
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544503 100 μL
PIMA544504 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544503 Supplier Invitrogen™ Supplier No. MA544503
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE tissue sections.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGRP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone BLR159J
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene Calca
Gene Accession No. P70160
Gene Alias alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6
Gene Symbols Calca
Host Species Rabbit
Immunogen Peptide representing a region between residue 75 and 125 of mouse Calcitonin gene-related Peptide 1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12310
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.