Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGRP1 Antibody (5), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP300518AF405
Description
CGRP1 Monoclonal antibody specifically detects CGRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
CGRP1 | |
Monoclonal | |
Alexa Fluor 405 | |
alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
Synthetic human alpha-CGRP | |
0.1 mL | |
Cardiovascular Biology, Cellular Signaling, Neuroscience, Signal Transduction | |
796 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
5 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction