Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CGS 21680 hydrochloride
GSA_VAGREENER_CHOICESDP

Catalog No. 106350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-635-0 50 mg
10-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-635-0 Supplier R&D Systems™ Supplier No. 1063/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. A2A agonist

CGS 21680 hydrochloride is a potent A2A adenosine receptor agonist (Ki = 27 nM, EC50 = 1.48-180 nM). CGS 21680 reduces venous resistance, increases heart rate and cardiac output in vivo. In an animal model of Huntington's disease, CGS21680 slows motor deterioration, prevents reduction in brain weight and reduces the size of mutant huntingtin aggregates and intranuclear inclusions. CGS21680 increases proliferation of neural progenitor cells, promotes survival of motoneurons and activates TrkB receptors. Also has anti-inflammatory effects in animal models of lung injury and reduces neurotoxicity induced by Kainate (Cat. No. 7065).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 124431-80-7
Molecular Formula C23H30ClN7O6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 535.99
Purity >98%
Molecular Weight (g/mol) 535.986
Chemical Name or Material CGS 21680 hydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.