Learn More
R&D Systems™ CGS 35066
GREENER_CHOICESDP
Catalog No. 251210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
25-121-0 10 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-121-0 Supplier R&D Systems™ Supplier No. 2512/10
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Endothelin-converting enzyme (ECE) inhibitor
CGS 35066 is a potent endothelin-converting enzyme (ECE) inhibitor that displays > 100-fold selectivity over neutral endopeptidase 24.11 (IC50 values are 22 and 2300 nM respectively). Blocks the hypertensive effects induced by big ET-1 in vitro and reduces the magnitude of cerebral vasospasm following subarachnoid hemorrhage (SAH).
Specifications
Quantity 10 mg
Product Line Tocris
Product Type CGS 35066
Molecular Weight (g/mol) 349.28
Inhibitors CGS 35066