Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CH25H Antibody [Janelia Fluor™ 585], Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320218JF585
Description
CH25H Polyclonal antibody specifically detects CH25H in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| CH25H | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9023 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 585 | |
| C25H, cholesterol 25-hydroxylase, Cholesterol 25-monooxygenase, EC 1.14.99.38, h25OH | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CH25H | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction