Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CH25H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32021850UL
Description
CH25H Polyclonal antibody specifically detects CH25H in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CH25H | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
C25H, cholesterol 25-hydroxylase, Cholesterol 25-monooxygenase, EC 1.14.99.38, h25OH | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CH25H | |
50 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
9023 | |
Store at 4&drg;C short term. Aliquot and store at -20&drg;C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction