Learn More
ChABC Antibody (1E10), Alexa Fluor™ 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP196141AF405
Catalog No. NBP196141W
Only null left
Add to Cart
Description
Description
ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
Monoclonal
Alexa Fluor 405
Chondroitinase ABC
Chondroitinase ABC
Protein G purified
RUO
Non-species specific
Purified
Western Blot, ELISA
10000000000
Western Blot, ELISA
Mouse
80 kDa
0.1 mL
Primary
Store at 4C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only