Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChABC Antibody (1E10), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196141AF594
Description
ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
ChABC | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Rat, Non-species specific | |
IgG |
Western Blot, ELISA | |
10000000000 | |
Western Blot, ELISA | |
Chondroitinase ABC | |
Chondroitinase ABC | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction