Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChABC Antibody (1E10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen ChABC
Clone 10000000000
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP196141SS
SDP
View Documents
Novus Biologicals
NBP196141SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP196141
SDP
View Documents
Novus Biologicals
NBP196141
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ChABC Monoclonal specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ChABC
1 mg/mL
Western Blot, ELISA
Unconjugated
Mouse
Neurodegeneration, Neuroscience
Chondroitinase ABC
IgG
Protein G purified
80 kDa
10000000000
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Monoclonal
Purified
RUO
Non-species specific
Chondroitinase ABC
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.