Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI-1 Antibody (LLBmi1-1) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$206.50 - $517.59

Specifications

Antigen BMI-1
Clone LLBmi1-1
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP196140SS
SDP
View Documents
Novus Biologicals
NBP196140SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP196140
SDP
View Documents
Novus Biologicals
NBP196140
0.1 mL
Each for $517.59
Only null left
Add to Cart
 
Description

Description

BMI-1 Monoclonal specifically detects BMI-1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow (Intracellular).
Specifications

Specifications

BMI-1
1.0 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Hematopoietic Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
P35226, P25916
648
Human Bmi1 protein [Swiss-Prot# P35226]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
LLBmi1-1
Western Blot 1:1000, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Monoclonal
Purified
RUO
Human, Mouse
B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4
BMI1
IgG2b κ
Protein G purified
42 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.