Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChABC Antibody (6A12), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196142R
Description
ChABC Monoclonal specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
ChABC | |
Monoclonal | |
DyLight 550 | |
Chondroitinase ABC | |
Chondroitinase ABC | |
Protein G purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, ELISA | |
6A12 | |
Western Blot, ELISA | |
Mouse | |
80 kDa | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction