Learn More
ChABC Antibody (6A12), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP196142IR
Catalog No. NBP196142IR
Only null left
Add to Cart
Description
Description
ChABC Monoclonal specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein G purified
Primary
Store at 4C in the dark.
IgG
Western Blot, ELISA
6A12
Western Blot, ELISA
Chondroitinase ABC
Chondroitinase ABC
0.1 mL
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.