Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChABC Antibody (6A12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196142MFV500
Description
ChABC Monoclonal antibody specifically detects ChABC in Nonspecies specific samples. It is validated for Western Blot, ELISASpecifications
ChABC | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, ELISA | |
6A12 | |
Western Blot, ELISA | |
Chondroitinase ABC | |
Chondroitinase ABC | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction