Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChABC Antibody (6A12), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ChABC |
---|---|
Clone | 6A12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP196142SS
![]() |
Novus Biologicals
NBP196142SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP196142
![]() |
Novus Biologicals
NBP196142 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
ChABC Monoclonal specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
ChABC | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Neurodegeneration, Neuroscience | |
Chondroitinase ABC | |
IgG | |
Protein G purified | |
80 kDa |
6A12 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Non-species specific | |
Chondroitinase ABC | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title