Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157685
Description
CHAC1 Polyclonal specifically detects CHAC1 in Human samples. It is validated for Western Blot.Specifications
| CHAC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cation transport regulator-like protein 1, ChaC, cation transport regulator homolog 1 (E. coli) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79094 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BUX1 | |
| CHAC1 | |
| Synthetic peptides corresponding to CHAC1(ChaC, cation transport regulator homolog 1 (E. coli)) The peptide sequence was selected from the C terminal of CHAC1. Peptide sequence TPQNPGYLGPAPEEAIATQILACRGFSGHNLEYLLRLADFMQLCGPQAQD. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction