Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHAC1 Antibody (OTI1E2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272191F
Description
CHAC1 Monoclonal specifically detects CHAC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CHAC1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CHAC1 | |
Full length recombinant protein of human CHAC1 (NP_077016) produced in HEK293T cells. | |
0.1 mL | |
79094.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E2 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
cation transport regulator-like protein 1, ChaC, cation transport regulator homolog 1 (E. coli) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction