Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHAC1 Antibody (OTI1E2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272191PE
Description
CHAC1 Monoclonal antibody specifically detects CHAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
CHAC1 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI1E2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
cation transport regulator-like protein 1, ChaC, cation transport regulator homolog 1 (E. coli) | |
Full length human recombinant protein of human CHAC1(NP_077016) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis, Cell Biology | |
79094 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction