Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAC1 Mouse anti-Human, Clone: OTI1E2, liquid, TrueMAB™
SDP

Catalog No. 501676932
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6932 Supplier OriGene Supplier No. TA507052
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.

Specifications

Antigen CHAC1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CHAC1
Gene Accession No. Q9BUX1
Gene Alias ChaC glutathione specific gamma-glutamylcyclotransferase 1
Gene Symbols CHAC1
Host Species Mouse
Immunogen Full length human recombit protein of human CHAC1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79094
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.