Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHAC1 Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA507010

Description
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.Specifications
CHAC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BUX1 | |
CHAC1 | |
Full length human recombit protein of human CHAC1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G1 | |
Unconjugated | |
CHAC1 | |
ChaC glutathione specific gamma-glutamylcyclotransferase 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79094 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction