Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578682 Supplier Invitrogen™ Supplier No. PA578682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, hESC, Neuro 2A, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12 Predicted Reactivity: Rat (90%), Xenopus laevis (84%), Chicken (83%), Rhesus Monkey (99%), Bovine (84%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CHAC2 catalyzes the cleavage glutathione into 5-oxoproline and a Cys-Gly dipeptide and acts specifically on glutathione, but not on other gamma-glutamyl peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHAC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Chac2
Gene Accession No. Q8WUX2, Q9CQG1
Gene Alias cation transport regulator-like protein 2; ChaC cation transport regulator homolog 2; ChaC, cation transport regulator 2; ChaC, cation transport regulator homolog 2; ChaC, cation transport regulator homolog 2 (E. coli); ChaC, cation transport regulator-like 2; Chac2; CHAC2 (HGNC:32363); Gamma-GCG 2; gamma-GCT acting on glutathione homolog 2; Glutathione-specific gamma-glutamylcyclotransferase 2; Putative glutathione-specific gamma-glutamylcyclotransferase 2; RGD1309120
Gene Symbols Chac2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CHAC2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494143, 68044
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.