Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114072 Supplier Invitrogen™ Supplier No. PA5114072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWVFGYGSLI WKVDFPYQDK LVGYITNYSR RFWQGSTDHR GVPGKPGRVV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 100%.

CHAC2 catalyzes the cleavage glutathione into 5-oxoproline and a Cys-Gly dipeptide and acts specifically on glutathione, but not on other gamma-glutamyl peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHAC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Chac2
Gene Accession No. Q8WUX2
Gene Alias cation transport regulator-like protein 2; ChaC cation transport regulator homolog 2; ChaC, cation transport regulator 2; ChaC, cation transport regulator homolog 2; ChaC, cation transport regulator homolog 2 (E. coli); ChaC, cation transport regulator-like 2; Chac2; CHAC2 (HGNC:32363); Gamma-GCG 2; gamma-GCT acting on glutathione homolog 2; Glutathione-specific gamma-glutamylcyclotransferase 2; Putative glutathione-specific gamma-glutamylcyclotransferase 2; RGD1309120
Gene Symbols Chac2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CHAC2 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.