Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553761
Description
Immunogen sequence: LRWLYLSENA LSSLQPGALD DVENLAKFHV DRNQLSSYPS AALSKLRVVE ELKLSHNPLK SIPDNAFQSF GRYLETLWLD NTNLEKFSDG AFLGVTT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
Specifications
CHAD | |
Polyclonal | |
Unconjugated | |
CHAD | |
Cartilage leucine-rich protein; Chad; Chondroadherin; chondroadherin proteoglycan; SLRR4A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1101 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15335 | |
CHAD | |
Recombinant protein corresponding to Human CHAD. Recombinant protein control fragment (Product #RP-91869). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction