Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572887
Description
CHAD Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
Specifications
CHAD | |
Polyclonal | |
Unconjugated | |
CHAD | |
Cartilage leucine-rich protein; Chad; Chondroadherin; chondroadherin proteoglycan; SLRR4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1101, 12643, 29195 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O15335, O55226, O70210 | |
CHAD | |
Chondroadherin antibody for a 17 amino acid peptide near the carboxy terminus of human Chondroadherin. The immunogen is within the last 50 amino acids of Chondroadherin. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction