Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHADL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552185 Supplier Invitrogen™ Supplier No. PA552185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEGAPRLGYL YLERNRFLQV PGAALRALPS LFSLHLQDNA VDRLAPGDLG RTRALRWVYL SGNRITEVSL GALGPARELE KLHLDRNQLR EVPTGALEGL PALLELQLSG NPLRALRDGA FQPVGRSL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

Potential negative modulator of chondrocyte differentiation. Inhibits collagen fibrillogenesis in vitro. May influence chondrocyte's differentiation by acting on its cellular collagenous microenvironment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHADL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHADL
Gene Accession No. Q6NUI6
Gene Alias AY100452; Chadl; chondroadherin like; chondroadherin pseudogene; chondroadherin-like; chondroadherin-like protein; chondroadherin-like protein-like; D930017K21Rik; leucine rich repeat containing 15 pseudogene; SLRR4B
Gene Symbols CHADL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHADL. Recombinant protein control fragment (Product #RP-91547).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150356
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.