Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Chaetocin
GREENER_CHOICESDP

Catalog No. 45041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-041 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-041 Supplier Tocris Bioscience™ Supplier No. 4504/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

SUV39H1 inhibitor

Histone methyltransferase SUV39H1 inhibitor (IC50 = 0.8 μM). Induces apoptosis in myeloma cell lines in vitro; exhibits antiproliferative activity in a mouse myeloma model in vivo.

Chemical Identifiers

CAS 28097-03-2
Molecular Formula C30H28N6O6S4

Specifications

Quantity 1 mg
Formula Weight 696.84
Purity >99%
Molecular Weight (g/mol) 696.83
Chemical Name or Material Chaetocin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.