Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAF1B Antibody (SS 24 1-68), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$243.50 - $341.50

Specifications

Antigen CHAF1B
Clone SS 24 1-68
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 0.1-1 ug
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB500211SS
SDP
View Documents
Novus Biologicals
NB500211SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB500211
SDP
View Documents
Novus Biologicals
NB500211
0.05 mL
Each for $341.50
Only null left
Add to Cart
 
Description

Description

CHAF1B Monoclonal specifically detects CHAF1B in Human, Mouse, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

CHAF1B
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 0.1-1 ug
Monoclonal
Ascites
RUO
Human, Mouse, Primate
Q13112
8208
Recombinant human CHAF1B [UniProt# Q13112]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
SS 24 1-68
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research
Ascites with 0.1% Sodium Azide
CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7
CHAF1B
IgG1
Unpurified
60 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.