Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHAF1B, Rabbit anti-Human, Clone: E17-L, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15984
Specifications
CHAF1B | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide). | |
CAF-1/CAF-IP60/CAF1/CAF1A/CAF1P60/MPHOSPH7/MPP7 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody |
Immunohistochemistry (Paraffin) | |
E17-L | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
chromatin assembly factor 1, subunit B (p60) | |
CHAF1B | |
A synthetic peptide corresponding to C-terminus of human CHAF1B. | |
RUO | |
8208 | |
Store at 4°C. Do not freeze. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction