Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chalcocite, naturally occurring mineral, grains, approximately, 0.06 to 0.19 in., Thermo Scientific™

Catalog No. p-7076428
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22205-45-4
Molecular Formula Cu2S
Molecular Weight (g/mol) 159.15
MDL Number MFCD00016062
InChI Key AQMRBJNRFUQADD-UHFFFAOYSA-N
Synonym copper i sulfide, cuprasulfide, copper sulfide cu2s, dicopper sulfide, dicopper sulphide, dicopper monosulfide, cuprous sulfide cu2s, unii-349m3c1rs1, cu2s, cupriosulfanyl copper
PubChem CID 62755
IUPAC Name bis(λ¹-copper(1+)) sulfanediide
SMILES [S--].[Cu+].[Cu+]

Specifications

Melting Point ∽1100°C
Quantity 1 g
Solubility Information Slightly soluble in hydrochloric acid,ammonium hydroxide. Insoluble in water.
Formula Weight 159.14
Packaging Glass bottle
Chemical Name or Material Chalcocite
EINECSNumber 244-842-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.